3-Amino-4-methylbenzonitrile 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD21733-10GEA
171
DKK
BLDPBD21733-10G
BLDPBD21733-1G
BLDPBD21733-5G
BLDPBD21733-25G
BLDPBD21733-100G
BLDPBD21733-500G
3-Amino-4-methylbenzonitrile 98%
3-Amino-4-methylbenzonitrile
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₈H₈N₂ Molekylvægt: 132,1600 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04039781 CAS-nummer: 60710-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...