4-(Ethoxycarbonyl)-2-nitrophenylboronic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD21583-10GEA 1120 DKK
BLDPBD21583-10G BLDPBD21583-5G BLDPBD21583-25G BLDPBD21583-1G BLDPBD21583-250MG
4-(Ethoxycarbonyl)-2-nitrophenylboronic acid 97%
4-(Ethoxycarbonyl)-2-nitrophenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₀BNO₆
Molekylvægt: 238,9898 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02179460
CAS-nummer: 5785-70-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR