2-Chloro-4-(trifluoromethyl)benzyl alcohol 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD21497-250MGEA
69.8
DKK
BLDPBD21497-250MG
BLDPBD21497-1G
BLDPBD21497-10G
BLDPBD21497-5G
BLDPBD21497-25G
2-Chloro-4-(trifluoromethyl)benzyl alcohol 97%
2-Chloro-4-(trifluoromethyl)benzyl alcohol
Storage condition: Sealed in dry, Room Temperature
Formel:
C₈H₆ClF₃O Molekylvægt: 210,58 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04972757 CAS-nummer: 56456-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...