3-(Methylsulphonyl)benzoic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD21276-5GEA 148 DKK
BLDPBD21276-5G BLDPBD21276-250MG BLDPBD21276-100G BLDPBD21276-25G BLDPBD21276-1G BLDPBD21276-10G
3-(Methylsulphonyl)benzoic acid 97%
3-(Methylsulphonyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈O₄S
Molekylvægt: 200,21 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00216488
CAS-nummer: 5345-27-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR