6-Chloro-2-methylnicotinic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD211877-25GEA 3140 DKK
BLDPBD211877-25G BLDPBD211877-250MG BLDPBD211877-5G BLDPBD211877-10G BLDPBD211877-100MG BLDPBD211877-1G
6-Chloro-2-methylnicotinic acid 98%
6-Chloro-2-methylnicotinic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₆ClNO₂
Molekylvægt: 171,581 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09842452
CAS-nummer: 137129-98-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR