Pyrazolo[1,5-a]pyrimidine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD211016-1GEA 528 DKK
BLDPBD211016-1G BLDPBD211016-250MG BLDPBD211016-25G BLDPBD211016-5G BLDPBD211016-10G
Pyrazolo[1,5-a]pyrimidine 98%
Pyrazolo[1,5-a]pyrimidine

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₅N₃
Molekylvægt: 119,124 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10699580
CAS-nummer: 274-71-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR