(2-Chloroethoxy)acetic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD210961-250MGEA 318 DKK
BLDPBD210961-250MG BLDPBD210961-100MG
(2-Chloroethoxy)acetic acid 95%
(2-Chloroethoxy)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₄H₇ClO₃
Molekylvægt: 138,55 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD17014832
CAS-nummer: 14869-41-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR