5-Amino-1-methyl-1,2-dihydropyridin-2-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD210918-25GEA 7690 DKK
BLDPBD210918-25G BLDPBD210918-1G BLDPBD210918-250MG BLDPBD210918-5G BLDPBD210918-100MG
5-Amino-1-methyl-1,2-dihydropyridin-2-one 95%
5-Amino-1-methyl-1,2-dihydropyridin-2-one

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₆H₈N₂O
Molekylvægt: 124,141 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09814480
CAS-nummer: 33630-96-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR