2-Methylpropane-1,2-diol 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD208742-10GEA 652 DKK
BLDPBD208742-10G BLDPBD208742-1G BLDPBD208742-25G BLDPBD208742-100G BLDPBD208742-5G
2-Methylpropane-1,2-diol 98%
2-Methylpropane-1,2-diol

Storage condition: Sealed in dry, Room Temperature

Formel: C₄H₁₀O₂
Molekylvægt: 90,121 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00971926
CAS-nummer: 558-43-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR