5-Chloro-2-(trifluoromethyl)aniline 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD20855-1GEA
225
DKK
BLDPBD20855-1G
BLDPBD20855-10G
BLDPBD20855-5G
5-Chloro-2-(trifluoromethyl)aniline 98%
5-Chloro-2-(trifluoromethyl)aniline
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₇H₅ClF₃N Molekylvægt: 195,5700 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08282771 CAS-nummer: 445-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...