6-Phenylpicolinic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD20641-1GEA 318 DKK
BLDPBD20641-1G BLDPBD20641-10G BLDPBD20641-5G BLDPBD20641-25G BLDPBD20641-250MG
6-Phenylpicolinic acid 95%
6-Phenylpicolinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₂H₉NO₂
Molekylvægt: 199,2054 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04114206
CAS-nummer: 39774-28-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR