4-Amino-α,α,α-trifluoro-o-toluic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD20609-5GEA 101 DKK
BLDPBD20609-5G BLDPBD20609-1G BLDPBD20609-25G BLDPBD20609-10G
4-Amino-α,α,α-trifluoro-o-toluic acid 97%
4-Amino-α,α,α-trifluoro-o-toluic acid
Formel: C₈H₆F₃NO₂
Molekylvægt: 205,14 g/mol
MDL Number: MFCD03407959
CAS-nummer: 393-06-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR