Methyl-5-chloropyrazine-2-carboxylate 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD20279-5GEA 125 DKK
BLDPBD20279-5G BLDPBD20279-25G BLDPBD20279-1G BLDPBD20279-10G BLDPBD20279-100G
Methyl-5-chloropyrazine-2-carboxylate 98%
Methyl-5-chloropyrazine-2-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₅ClN₂O₂
Molekylvægt: 172,57 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01632102
CAS-nummer: 33332-25-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR