N-Boc-D-alaninol 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD20223-25GEA 272 DKK
BLDPBD20223-25G BLDPBD20223-5G BLDPBD20223-100G BLDPBD20223-500G BLDPBD20223-10G
N-Boc-D-alaninol 97%
N-Boc-D-alaninol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₇NO₃
Molekylvægt: 175,2 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00235912
CAS-nummer: 106391-86-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR