1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD200042-250MGEA 140 DKK
BLDPBD200042-250MG BLDPBD200042-5G BLDPBD200042-1G
1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride 98%
1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₉ClFNO
Molekylvægt: 177,6039 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01114584
CAS-nummer: 215599-91-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR