(2-Chloro-1,3-thiazol-5-yl)methyl phenyl ether 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD199713-250MGEA 249 DKK
BLDPBD199713-250MG BLDPBD199713-1G BLDPBD199713-100MG BLDPBD199713-5G
(2-Chloro-1,3-thiazol-5-yl)methyl phenyl ether 95+%
(2-Chloro-1,3-thiazol-5-yl)methyl phenyl ether

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₈NOClS
Molekylvægt: 225,6946 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01443853
CAS-nummer: 339018-23-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR