(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD19780-100MGEA 256 DKK
BLDPBD19780-100MG BLDPBD19780-250MG BLDPBD19780-5G BLDPBD19780-1G
(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid 97%
(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₈BrNO₄
Molekylvægt: 344,20 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01320860
CAS-nummer: 261380-20-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR