3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD196712-1GEA
690
DKK
BLDPBD196712-1G
BLDPBD196712-250MG
BLDPBD196712-100MG
BLDPBD196712-5G
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile 98%
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₈H₄BrN₃ Molekylvægt: 222,05 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06659617 CAS-nummer: 885950-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...