Ethyl-2,5-dichloroisonicotinate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD196639-250MGEA 287 DKK
BLDPBD196639-250MG
Ethyl-2,5-dichloroisonicotinate 95%
Ethyl-2,5-dichloroisonicotinate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₇Cl₂NO₂
Molekylvægt: 220,0500 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09972252
CAS-nummer: 603122-76-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR