4-Mercaptophenylboronic acid 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BLDPBD19568-5GEA
838
DKK
BLDPBD19568-5G
BLDPBD19568-1G
BLDPBD19568-25G
4-Mercaptophenylboronic acid 95%
4-Mercaptophenylboronic acid
Formel:
C₆H₇BO₂S Molekylvægt: 153,9946 g/mol |
MDL Number:
MFCD03093887 CAS-nummer: 237429-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...