3-Cyano-4-fluorophenylboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD19364-5GEA 388 DKK
BLDPBD19364-5G BLDPBD19364-10G BLDPBD19364-100G BLDPBD19364-250MG BLDPBD19364-25G BLDPBD19364-100MG BLDPBD19364-1G
3-Cyano-4-fluorophenylboronic acid 98%
3-Cyano-4-fluorophenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₅BFNO₂
Molekylvægt: 164,93 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03095130
CAS-nummer: 214210-21-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR