6-Bromoanthranilic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD19304-5GEA 187 DKK
BLDPBD19304-5G BLDPBD19304-10G BLDPBD19304-1G BLDPBD19304-25G BLDPBD19304-100G BLDPBD19304-0.025G
6-Bromoanthranilic acid 98%
6-Bromoanthranilic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₆BrNO₂
Molekylvægt: 216,03 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03618455
CAS-nummer: 20776-48-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR