1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD18896-250MGEA
125
DKK
BLDPBD18896-250MG
1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid 95%
1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Storage condition: Sealed in dry, Room Temperature
Formel:
C₉H₁₄N₂O₂ Molekylvægt: 182,22 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00084917 CAS-nummer: 175277-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...