4-Ethylcaprylic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD18745-25GEA 869 DKK
BLDPBD18745-25G BLDPBD18745-10G BLDPBD18745-1G BLDPBD18745-5G BLDPBD18745-100G
4-Ethylcaprylic acid 98%
4-Ethylcaprylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₀O₂
Molekylvægt: 172,27 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00506494
CAS-nummer: 16493-80-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR