Cinnamyltriphenylphosphonium bromide 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD1861-1GEA 85.3 DKK
BLDPBD1861-1G BLDPBD1861-5G BLDPBD1861-25G
Cinnamyltriphenylphosphonium bromide 97%
Cinnamyltriphenylphosphonium bromide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₇H₂₄BrP
Molekylvægt: 459,3600 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00040767
CAS-nummer: 7310-74-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR