4,6-Dichloro-5-(2-methoxyphenoxy)-2,2-bipyrimidine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD18501-5GEA 349 DKK
BLDPBD18501-5G BLDPBD18501-25G BLDPBD18501-1G BLDPBD18501-100G BLDPBD18501-10G
4,6-Dichloro-5-(2-methoxyphenoxy)-2,2-bipyrimidine 98%
4,6-Dichloro-5-(2-methoxyphenoxy)-2,2-bipyrimidine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₅H₁₀Cl₂N₄O₂
Molekylvægt: 349,17 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 150728-13-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR