(S)-1-Boc-pyrrolidine-3-carboxylic acid 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD18262-5GEA
1120
DKK
BLDPBD18262-5G
BLDPBD18262-1G
BLDPBD18262-25G
BLDPBD18262-100G
BLDPBD18262-10G
BLDPBD18262-100MG
BLDPBD18262-250MG
(S)-1-Boc-pyrrolidine-3-carboxylic acid 97%
(S)-1-Boc-pyrrolidine-3-carboxylic acid
Storage condition: Sealed in dry, 2-8°C
Formel:
C₁₀H₁₇NO₄ Molekylvægt: 215,25 g/mol Opbevaringstemperatur: Køleskab |
CAS-nummer:
140148-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...