1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD182473-5GEA 690 DKK
BLDPBD182473-5G BLDPBD182473-1G BLDPBD182473-25G
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride 97%
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₉ClFNO
Molekylvægt: 177,6039 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01114581
CAS-nummer: 51572-89-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR