N-Methyl-3-pyrrolidinol 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD18026-25GEA 900 DKK
BLDPBD18026-25G BLDPBD18026-10G BLDPBD18026-100G BLDPBD18026-5G BLDPBD18026-1G
N-Methyl-3-pyrrolidinol 97%
N-Methyl-3-pyrrolidinol

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₁₁NO
Molekylvægt: 101,15 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003176
CAS-nummer: 13220-33-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR