Sirtinol 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD1800-1MGEA 171 DKK
BLDPBD1800-1MG
Sirtinol 98%
Sirtinol

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₂₆H₂₂N₂O₂
Molekylvægt: 394,46 g/mol
Opbevaringstemperatur: Fryser
CAS-nummer: 410536-97-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR