4-(Methylcarbamoyl)phenylboronic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD17790-250MGEA 46.6 DKK
BLDPBD17790-250MG BLDPBD17790-5G BLDPBD17790-100G BLDPBD17790-25G BLDPBD17790-10G BLDPBD17790-0.025G BLDPBD17790-1G
4-(Methylcarbamoyl)phenylboronic acid 95%
4-(Methylcarbamoyl)phenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₈H₁₀BNO₃
Molekylvægt: 178,98 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03788427
CAS-nummer: 121177-82-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR