Ethyl-2-chloro-6-methylisonicotinate 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD174192-5GEA 724 DKK
BLDPBD174192-5G BLDPBD174192-250MG BLDPBD174192-1G
Ethyl-2-chloro-6-methylisonicotinate 97%
Ethyl-2-chloro-6-methylisonicotinate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₀ClNO₂
Molekylvægt: 199,64 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09842151
CAS-nummer: 3998-88-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR