3-Amino-6,7-difluoro-1H-indazole 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD173911-5GEA 1800 DKK
BLDPBD173911-5G BLDPBD173911-1G BLDPBD173911-250MG
3-Amino-6,7-difluoro-1H-indazole 95%
3-Amino-6,7-difluoro-1H-indazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₅F₂N₃
Molekylvægt: 169,1300 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD13191632
CAS-nummer: 706805-37-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR