4-(Isopropylamino)diphenylamine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD17289-25GEA 54.3 DKK
BLDPBD17289-25G
4-(Isopropylamino)diphenylamine 98%
4-(Isopropylamino)diphenylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₅H₁₈N₂
Molekylvægt: 226,32 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00026348
CAS-nummer: 101-72-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR