2-Amino-N-methyl-N-phenylbenzamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD172694-250MGEA 652 DKK
BLDPBD172694-250MG
2-Amino-N-methyl-N-phenylbenzamide 95%
2-Amino-N-methyl-N-phenylbenzamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₄H₁₄N₂O
Molekylvægt: 226,2738 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00443942
CAS-nummer: 6632-37-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR