Cyclohexyldiphenylphosphine oxide 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD16928-1GEA 69.8 DKK
BLDPBD16928-1G BLDPBD16928-0.025G BLDPBD16928-25G BLDPBD16928-10G BLDPBD16928-5G BLDPBD16928-100G BLDPBD16928-250MG
Cyclohexyldiphenylphosphine oxide 98%
Cyclohexyldiphenylphosphine oxide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₈H₂₁OP
Molekylvægt: 284,34 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014301
CAS-nummer: 13689-20-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR