3-Bromo-5-fluoro-1H-indazole 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD168072-100MGEA 318 DKK
BLDPBD168072-100MG BLDPBD168072-1G BLDPBD168072-250MG
3-Bromo-5-fluoro-1H-indazole 95%
3-Bromo-5-fluoro-1H-indazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₄BrFN₂
Molekylvægt: 215,0200 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07781639
CAS-nummer: 885519-08-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR