9-Phenylcarbazole 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD16804-25GEA 187 DKK
BLDPBD16804-25G BLDPBD16804-10G BLDPBD16804-100G BLDPBD16804-5G
9-Phenylcarbazole 98%
9-Phenylcarbazole

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₈H₁₃N
Molekylvægt: 243,30 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004965
CAS-nummer: 1150-62-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR