7-Isoquinolinecarboxylic acid 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD167889-100MGEA 442 DKK
BLDPBD167889-100MG BLDPBD167889-250MG
7-Isoquinolinecarboxylic acid 95%
7-Isoquinolinecarboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₇NO₂
Molekylvægt: 173,1681 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10699235
CAS-nummer: 221050-96-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR