Isoquinoline-6-carboxaldehyde 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD167856-100MGEA 1070 DKK
BLDPBD167856-100MG BLDPBD167856-250MG
Isoquinoline-6-carboxaldehyde 95%
Isoquinoline-6-carboxaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₇NO
Molekylvægt: 157,169 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06227438
CAS-nummer: 173089-81-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR