6-Ethoxy-4-hydroxyquinoline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD167646-250MGEA 225 DKK
BLDPBD167646-250MG BLDPBD167646-100MG BLDPBD167646-1G BLDPBD167646-5G
6-Ethoxy-4-hydroxyquinoline 95%
6-Ethoxy-4-hydroxyquinoline

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₁NO₂
Molekylvægt: 189,211 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02332227
CAS-nummer: 303121-11-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR