(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD1670-100GEA
1550
DKK
BLDPBD1670-100G
BLDPBD1670-10G
BLDPBD1670-5G
BLDPBD1670-25G
BLDPBD1670-1G
(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide 97%
(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₂₄H₂₆BrO₂P Molekylvægt: 457,35 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00075527 CAS-nummer: 59159-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...