Methyl-5-bromo-2-iodobenzoate 96%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD161272-100GEA
1350
DKK
BLDPBD161272-100G
BLDPBD161272-10G
BLDPBD161272-25G
BLDPBD161272-1G
BLDPBD161272-500G
BLDPBD161272-5G
Methyl-5-bromo-2-iodobenzoate 96%
Methyl-5-bromo-2-iodobenzoate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₈H₆BrIO₂ Molekylvægt: 340,9400 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00144771 CAS-nummer: 181765-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...