2-Bromo-4-fluoro-6-nitrotoluene 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD161239-5GEA
264
DKK
BLDPBD161239-5G
BLDPBD161239-1G
BLDPBD161239-10G
BLDPBD161239-100G
BLDPBD161239-250MG
BLDPBD161239-25G
2-Bromo-4-fluoro-6-nitrotoluene 97%
2-Bromo-4-fluoro-6-nitrotoluene
Storage condition: Sealed in dry, Room Temperature
Formel:
C₇H₅BrFNO₂ Molekylvægt: 234,0225 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03094189 CAS-nummer: 502496-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...