4-Cyclopropylaniline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD160279-250MGEA 349 DKK
BLDPBD160279-250MG BLDPBD160279-1G
4-Cyclopropylaniline 95%
4-Cyclopropylaniline
Formel: C₉H₁₁N
Molekylvægt: 133,19 g/mol
MDL Number: MFCD00019219
CAS-nummer: 3158-71-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR