3-Brom-2-nitroanisol 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD160153-1GEA 101 DKK
BLDPBD160153-1G BLDPBD160153-250MG
3-Brom-2-nitroanisol 97%
3-Brom-2-nitroanisol

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₆BrNO₃
Molekylvægt: 232,0314 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11101044
CAS-nummer: 500298-30-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR