2-Carboxy-4-fluorophenylboronic acid 95%
Leverandør: BLD PHARMATECH GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD158317-5GEA
613
DKK
BLDPBD158317-5G
BLDPBD158317-1G
BLDPBD158317-25G
2-Carboxy-4-fluorophenylboronic acid 95%
2-Carboxy-4-fluorophenylboronic acid
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₇H₆BFO₄ Molekylvægt: 183,9296 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD08235078 CAS-nummer: 874290-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...