4-Fluoro-2-methoxyaniline 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD157692-25GEA
287
DKK
BLDPBD157692-25G
BLDPBD157692-100G
BLDPBD157692-500G
BLDPBD157692-5G
BLDPBD157692-1G
4-Fluoro-2-methoxyaniline 98%
4-Fluoro-2-methoxyaniline
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₇H₈FNO Molekylvægt: 141,14 g/mol Opbevaringstemperatur: Stue temperatur |
CAS-nummer:
450-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...