4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD157659-10GEA
2900
DKK
BLDPBD157659-10G
BLDPBD157659-250MG
BLDPBD157659-100MG
BLDPBD157659-1G
BLDPBD157659-5G
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 97%
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₁₆H₂₃BO₄ Molekylvægt: 290,1624 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD08704684 CAS-nummer: 859169-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...