4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD15623-1GEA 1620 DKK
BLDPBD15623-1G BLDPBD15623-100MG BLDPBD15623-250MG BLDPBD15623-5G
4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%
4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane)

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₄H₂₁NO₅
Molekylvægt: 283,3202 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00068686
CAS-nummer: 60835-71-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR